DB-5 Columns, Agilent Technologies
DB-5 Columns, Agilent Technologies
Certificates
DB-5 Columns, Agilent Technologies
79 Options Available Below
Product Details & Documents
High-quality, nonpolar, general-purpose columns with low bleed and a high temperature limit.
  • (5%-Phenyl)-methylpolysiloxane
  • Nonpolar, low bleed and with a high temperature limit
  • Bonded, crosslinked, and solvent rinsible
  • Wide range of column dimensions available
  • Excellent general purpose column with a wide range of applications
  • Equivalent to USP Phase G27
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-5 is nonpolar and low bleed and is available in a range of column dimensions. DB-5 is also bonded, crosslinked, and solvent-rinsible, and has a high temperature limit. It is equivalent to USP phase G27. The DB-5 column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA127-5013
  • CA127-501E
  • CAAG127-5012
  • CAAG127-501N
  • CAAG127-5022
  • CAAG127-5023
  • CA121-5012
  • CA121-5022
  • CA121-5042
  • AG121-5012E
  • CAAG128-5012
  • CAAG128-5022
  • CAAG1225002LTM
  • CA122-5012
  • CA122-5013
  • CAG3900-65006
  • CAAG122-5011
  • CAAG122-501E
  • CAAG122-5022
  • CA122-5032
  • CA122-5033
  • CA122-503E
  • CA122-5631
  • CA122-5632
  • CAAG122-5031
  • CAAG122-5032E
  • CAAG1225032INT
  • CAAG1225033LTM
  • CAAG122-5052
  • CA122-5061
  • CA122-5062
  • CA122-5063
  • CAAG122-5063E
  • CA123-500ELTM
  • CAAG123-500E
  • CA123-5012
  • CA123-5013
  • CAAG123-5011
  • CAAG123-5013E
  • CAAG1235012LTM
  • CAAG123-5022
  • CAAG123-5026
  • CAAG123-502D
  • CA123-5032
  • CA123-5032E
  • CA123-5033
  • CA123-503B
  • CA123-503E
  • CAAG123-5031
  • CAAG123-5033E
  • CAAG123-5034
  • CAAG1235032INT
  • CAAG123503ELTM
  • CAAG1235003LTM
  • CA123-5056
  • CAAG123-5053
  • CAAG123-5062
  • CAAG123-5063
  • CAAG124-5032
  • CAAG124-5037
  • CAAG125-50HB
  • CA125-5012
  • CAAG125-5017
  • CAAG125-501J
  • CAAG125-501K
  • CAAG125-5025
  • CA125-5032
  • CA125-5035
  • CA125-503J
  • CAAG125-5032E
  • CAAG125-5034
  • CAAG125-5035E
  • CAAG125-5037
  • CAAG125-503B
  • CAAG125-503K
  • CA125-5065
  • CAAG125-5062
  • CAAG125-5065E
  • CAAG125-503D
  • Polarity:
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • USP code:
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Type de colonne
    Phase
    Column Hardware
    Épaisseur du film
    Ø int.
    Longueur
    Availability
    Price
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.40 µm
    0.10 mm
    10 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $630.00
    $630.00
    Each
       $630.00
    Supplier Number: 127-5013
    Spécifications:
    More
    Specifications
    Cat. No.
    CA127-5013
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5
    Fused silica
    0.17 µm
    0.10 mm
    10 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $630.00
    $630.00
    Each
       $630.00
    Supplier Number: 127-501E
    Spécifications:
    More
    Specifications
    Cat. No.
    CA127-501E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.1 µm
    0.10 mm
    10 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $693.00
    $693.00
    Each
       $693.00
    Supplier Number: 127-5012
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG127-5012
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.33 µm
    0.10 mm
    10 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $693.00
    $693.00
    Each
       $693.00
    Supplier Number: 127-501N
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG127-501N
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.1 µm
    0.10 mm
    20 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,080.00
    $1,080.00
    Each
       $1,080.00
    Supplier Number: 127-5022
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG127-5022
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.40 µm
    0.10 mm
    20 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,080.00
    $1,080.00
    Each
       $1,080.00
    Supplier Number: 127-5023
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG127-5023
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.18 µm
    0.18 mm
    10 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $505.00
    $505.00
    Each
       $505.00
    Supplier Number: 121-5012
    Spécifications:
    More
    Specifications
    Cat. No.
    CA121-5012
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.18 µm
    0.18 mm
    20 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $802.00
    $802.00
    Each
       $802.00
    Supplier Number: 121-5022
    Spécifications:
    More
    Specifications
    Cat. No.
    CA121-5022
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.18 µm
    0.18 mm
    40 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,490.00
    $1,490.00
    Each
       $1,490.00
    Supplier Number: 121-5042
    Spécifications:
    More
    Specifications
    Cat. No.
    CA121-5042
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC Column, 5 inch cage
    Capillaire
    DB-5
    Fused silica
    0.18 µm
    0.18 mm
    10 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $644.00
    $644.00
    Each
       $644.00
    Supplier Number: 121-5012E
    Spécifications:
    More
    Specifications
    Cat. No.
    AG121-5012E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.33 µm
    0.20 mm
    12 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $512.00
    $512.00
    Each
       $512.00
    Supplier Number: 128-5012
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG128-5012
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.33 µm
    0.20 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $871.00
    $871.00
    Each
       $871.00
    Supplier Number: 128-5022
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG128-5022
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • LTM column module
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.25 mm
    10 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,140.00
    $2,140.00
    Each
       $2,140.00
    Supplier Number: 122-5002LTM
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG1225002LTM
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.25 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $558.00
    $558.00
    Each
       $558.00
    Supplier Number: 122-5012
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5012
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.25 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $558.00
    $558.00
    Each
       $558.00
    Supplier Number: 122-5013
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5013
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC Column, 7 inch cage
    Capillaire
    DB-CSI
    Fused silica
    0.25 µm
    0.25 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,870.00
    $1,870.00
    Each
       $1,870.00
    Supplier Number: G3900-65006
    Spécifications:
    More
    Specifications
    Cat. No.
    CAG3900-65006
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.1 µm
    0.25 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $614.00
    $614.00
    Each
       $614.00
    Supplier Number: 122-5011
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5011
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.50 µm
    0.25 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $614.00
    $614.00
    Each
       $614.00
    Supplier Number: 122-501E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-501E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.25 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $961.00
    $961.00
    Each
       $961.00
    Supplier Number: 122-5022
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5022
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $937.00
    $937.00
    Each
       $937.00
    Supplier Number: 122-5032
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5032
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $937.00
    $937.00
    Each
       $937.00
    Supplier Number: 122-5033
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5033
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5
    Fused silica
    0.50 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $937.00
    $937.00
    Each
       $937.00
    Supplier Number: 122-503E
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-503E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5.625
    Fused silica
    0.25 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $946.00
    $946.00
    Each
       $946.00
    Supplier Number: 122-5631
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5631
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC Column, 7 inch cage
    Capillaire
    DB-5.625
    Fused silica
    0.50 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $946.00
    $946.00
    Each
       $946.00
    Supplier Number: 122-5632
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5632
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.1 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,040.00
    $1,040.00
    Each
       $1,040.00
    Supplier Number: 122-5031
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5031
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,160.00
    $1,160.00
    Each
       $1,160.00
    Supplier Number: 122-5032E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5032E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column module
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,200.00
    $1,200.00
    Each
       $1,200.00
    Supplier Number: 122-5032-INT
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG1225032INT
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • LTM column module
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.25 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,580.00
    $2,580.00
    Each
       $2,580.00
    Supplier Number: 122-5033LTM
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG1225033LTM
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.25 mm
    50 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,730.00
    $1,730.00
    Each
       $1,730.00
    Supplier Number: 122-5052
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5052
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.10 µm
    0.25 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,650.00
    $1,650.00
    Each
       $1,650.00
    Supplier Number: 122-5061
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5061
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.25 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,650.00
    $1,650.00
    Each
       $1,650.00
    Supplier Number: 122-5062
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5062
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.25 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,650.00
    $1,650.00
    Each
       $1,650.00
    Supplier Number: 122-5063
    Spécifications:
    More
    Specifications
    Cat. No.
    CA122-5063
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.25 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,030.00
    $2,030.00
    Each
       $2,030.00
    Supplier Number: 122-5063E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG122-5063E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.50 µm
    0.32 mm
    10 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,100.00
    $2,100.00
    Each
       $2,100.00
    Supplier Number: 123-500ELTM
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-500ELTM
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.50 µm
    0.32 mm
    10 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $586.00
    $586.00
    Each
       $586.00
    Supplier Number: 123-500E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-500E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.32 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $605.00
    $605.00
    Each
       $605.00
    Supplier Number: 123-5012
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-5012
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.32 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $605.00
    $605.00
    Each
       $605.00
    Supplier Number: 123-5013
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-5013
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.1 µm
    0.32 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $666.00
    $666.00
    Each
       $666.00
    Supplier Number: 123-5011
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5011
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.32 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $746.00
    $746.00
    Each
       $746.00
    Supplier Number: 123-5013E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5013E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • LTM column module
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.32 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,190.00
    $2,190.00
    Each
       $2,190.00
    Supplier Number: 123-5012LTM
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG1235012LTM
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.32 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $987.00
    $987.00
    Each
       $987.00
    Supplier Number: 123-5022
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5022
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.52 µm
    0.32 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $987.00
    $987.00
    Each
       $987.00
    Supplier Number: 123-5026
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5026
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.17 µm
    0.32 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $987.00
    $987.00
    Each
       $987.00
    Supplier Number: 123-502D
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-502D
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,020.00
    $1,020.00
    Each
       $1,020.00
    Supplier Number: 123-5032
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-5032
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,140.00
    $1,140.00
    Each
       $1,140.00
    Supplier Number: 123-5032E
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-5032E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,020.00
    $1,020.00
    Each
       $1,020.00
    Supplier Number: 123-5033
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-5033
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.50 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,020.00
    $1,020.00
    Each
       $1,020.00
    Supplier Number: 123-503B
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-503B
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5
    Fused silica
    0.50 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,020.00
    $1,020.00
    Each
       $1,020.00
    Supplier Number: 123-503E
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-503E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.1 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,120.00
    $1,120.00
    Each
       $1,120.00
    Supplier Number: 123-5031
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5031
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,250.00
    $1,250.00
    Each
       $1,250.00
    Supplier Number: 123-5033E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5033E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    3.00 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,020.00
    $1,020.00
    Each
       $1,020.00
    Supplier Number: 123-5034
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5034
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column module
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,300.00
    $1,300.00
    Each
       $1,300.00
    Supplier Number: 123-5032-INT
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG1235032INT
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • LTM column module
    Capillaire
    DB-5
    Fused silica
    0.50 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,670.00
    $2,670.00
    Each
       $2,670.00
    Supplier Number: 123-503ELTM
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123503ELTM
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • LTM column module
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.32 mm
    5 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,780.00
    $1,780.00
    Each
       $1,780.00
    Supplier Number: 123-5003LTM
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG1235003LTM
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.52 µm
    0.32 mm
    50 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,610.00
    $1,610.00
    Each
       $1,610.00
    Supplier Number: 123-5056
    Spécifications:
    More
    Specifications
    Cat. No.
    CA123-5056
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.32 mm
    50 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,770.00
    $1,770.00
    Each
       $1,770.00
    Supplier Number: 123-5053
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5053
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.32 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,970.00
    $1,970.00
    Each
       $1,970.00
    Supplier Number: 123-5062
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5062
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.32 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,970.00
    $1,970.00
    Each
       $1,970.00
    Supplier Number: 123-5063
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG123-5063
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.27 µm
    0.45 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,240.00
    $1,240.00
    Each
       $1,240.00
    Supplier Number: 124-5032
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG124-5032
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.42 µm
    0.45 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,240.00
    $1,240.00
    Each
       $1,240.00
    Supplier Number: 124-5037
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG124-5037
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    2.65 µm
    0.53 mm
    10 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $566.00
    $566.00
    Each
       $566.00
    Supplier Number: 125-50HB
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-50HB
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.50 µm
    0.53 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $618.00
    $618.00
    Each
       $618.00
    Supplier Number: 125-5012
    Spécifications:
    More
    Specifications
    Cat. No.
    CA125-5012
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.50 µm
    0.53 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $680.00
    $680.00
    Each
       $680.00
    Supplier Number: 125-5017
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-5017
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.53 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $680.00
    $680.00
    Each
       $680.00
    Supplier Number: 125-501J
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-501J
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.53 mm
    15 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $680.00
    $680.00
    Each
       $680.00
    Supplier Number: 125-501K
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-501K
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    5.00 µm
    0.53 mm
    25 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,130.00
    $1,130.00
    Each
       $1,130.00
    Supplier Number: 125-5025
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-5025
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.50 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,130.00
    $1,130.00
    Each
       $1,130.00
    Supplier Number: 125-5032
    Spécifications:
    More
    Specifications
    Cat. No.
    CA125-5032
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    5.00 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,130.00
    $1,130.00
    Each
       $1,130.00
    Supplier Number: 125-5035
    Spécifications:
    More
    Specifications
    Cat. No.
    CA125-5035
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.00 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,130.00
    $1,130.00
    Each
       $1,130.00
    Supplier Number: 125-503J
    Spécifications:
    More
    Specifications
    Cat. No.
    CA125-503J
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5
    Fused silica
    1.50 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,390.00
    $1,390.00
    Each
       $1,390.00
    Supplier Number: 125-5032E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-5032E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    3.00 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,240.00
    $1,240.00
    Each
       $1,240.00
    Supplier Number: 125-5034
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-5034
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5
    Fused silica
    5.00 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,390.00
    $1,390.00
    Each
       $1,390.00
    Supplier Number: 125-5035E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-5035E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.50 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,240.00
    $1,240.00
    Each
       $1,240.00
    Supplier Number: 125-5037
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-5037
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    2.65 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,240.00
    $1,240.00
    Each
       $1,240.00
    Supplier Number: 125-503B
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-503B
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.25 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,240.00
    $1,240.00
    Each
       $1,240.00
    Supplier Number: 125-503K
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-503K
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    5.00 µm
    0.53 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,810.00
    $1,810.00
    Each
       $1,810.00
    Supplier Number: 125-5065
    Spécifications:
    More
    Specifications
    Cat. No.
    CA125-5065
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    1.50 µm
    0.53 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,990.00
    $1,990.00
    Each
       $1,990.00
    Supplier Number: 125-5062
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-5062
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 5" cage
    Capillaire
    DB-5
    Fused silica
    5.00 µm
    0.53 mm
    60 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,230.00
    $2,230.00
    Each
       $2,230.00
    Supplier Number: 125-5065E
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-5065E
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41
  • GC column, 7" cage
    Capillaire
    DB-5
    Fused silica
    0.88 µm
    0.53 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,240.00
    $1,240.00
    Each
       $1,240.00
    Supplier Number: 125-503D
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG125-503D
    Polarity
    Low-polarity stationary phase
    USP code
    G27, G36, G41