J&W Ultra Inert DB-35ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Improved inertness and exceptionally low bleed, with great peak shapes for challenging active analytes such as pesticides and drugs of abuse.
- Individually tested with the Ultra Inert DB-35ms test probe mixture
- Consistent column inertness performance
- Exceptionally low column bleed
- Great peak shapes for challenging active analytes, with excellent signal-to-noise ratios
- Minimum compound adsorption or degradation
- Supports 0.18 mm int.Ø column configuration for higher sample throughput
Agilent J&W DB-35ms Ultra Inert is a mid-polarity column for applications where different selectivity is needed for enhanced separations, including pesticide analysis and drugs of abuse testing. This column quantifies components at a lower level, with broader linearity and optimized separation, ultimately increasing productivity.
Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency.
Specifications
- Catalog No:
- CA122-3812UI
- CAAG1223832UNT
- CAAG123-3832UI
- CA122-3832UI
- CAAG1223812UNT
- CAAG121-3822UI
- CAAG1233832UNT
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- USP code:
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42
- G28, G32, G42