J&W 8890 GC Columns with Smart Key, Agilent Technologies
Various GC columns with a Smart Key included, made specifically to fit the 8890 GC.
- Smart Key allows self-configuration, usage monitoring, and help automating method development when using 8890 GC
- Wide selection of available stationary phases (low, mid, and high polarity, application-specific stationary phases)
- Strict quality control specifications and testing ensure long-term column-to-column performance and accurate separations
- Optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials, and gas clean filters
- For use with the Agilent 8890 GC system
Agilent J&W offers a selection of GC columns with Smart Key (compatible only with the 8890 GC). Smart Keys allow for immediate identification and usage monitoring of your GC column. A variety of stationary phases are available, ranging from Ultra Inert and Poly Ethylene Glycol (PEG) to unique, application-specific stationary phases. If your configuration is not available, please request a custom column with Smart Key through the Custom Column Shop.
- Catalog No:
- CAAG1225532UIK
- CAAG1225532KEY
- CAAG19091S433K
- CAAG19091S43UK
- CAAG1257032KEY
- CAAG1225032KEY
- CAAG1227032KEY
- CAAGCP8944-KEY
- CAAG19091J413K
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- High-polarity stationary phase
- Low-polarity stationary phase
- High-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
Frequently Bought Together