ContactUsLinkComponent
VWR® Eco Premium ULT Freezers
New Product
undefined
VWR® Eco Premium ULT Freezers
New Product
  • Catalog No:
  • 76533-506
  • 76683-450
  • MP76533-506
  • MP76683-450
  • MP76514-176
  • MP76533-504
  • 76514-174
  • 76514-176
  • MP76514-174
  • Housing W×D×H:
  • 1110×938×1966 mm (44×37×77")
  • 706×938×1966 mm (28×37×77")
  • 1110×938×1966 mm (44×37×77")
  • 706×938×1966 mm (28×37×77")
  • 1110×938×1966 mm (44×37×77")
  • 826×938×1966 mm (33×37×77")
  • 826×938×1966 mm (33×37×77")
  • 1110×938×1966 mm (44×37×77")
  • 826×938×1966 mm (33×37×77")
  • Model:
  • VWR ULT Freezer 528 Eco Premium
  • VWR ULT freezer 264 Eco Premium
  • VWR ULT Freezer 528 Eco Premium
  • VWR ULT freezer 264 Eco Premium
  • VWR ULT Freezer 528 Eco Premium
  • VWR ULT Freezer 352 Eco Premium
  • VWR ULT Freezer 352 Eco Premium
  • VWR ULT Freezer 528 Eco Premium
  • VWR ULT Freezer 352 Eco Premium
  • Environmentally Preferable:
  • Low Manufacturing Impact, Energy Efficient
  • Low Manufacturing Impact, Energy Efficient
  • Low Manufacturing Impact, Energy Efficient
  • Low Manufacturing Impact, Energy Efficient
  • Low Manufacturing Impact, Energy Efficient
  • Low Manufacturing Impact, Energy Efficient
  • Low Manufacturing Impact, Energy Efficient
  • Low Manufacturing Impact, Energy Efficient
  • Low Manufacturing Impact, Energy Efficient
  • Cat. No.:
  • 76533-506
  • 76683-450
  • 76533-506
  • 76683-450
  • 76514-176
  • 76533-504
  • 76514-174
  • 76514-176
  • 76514-174
  • Electrical:
  • 208 V, 60 Hz
  • 208 V, 60 Hz
  • 208 V, 60 Hz
  • 208 V, 60 Hz
  • 120 V, 60 Hz
  • 208 V, 60 Hz
  • 120 V, 60 Hz
  • 120 V, 60 Hz
  • 120 V, 60 Hz
  • Noise level:
  • 47 db(A)
  • 47 db(A)
  • 47 db(A)
  • 47 db(A)
  • 47 db(A)
  • 47 db(A)
  • 47 db(A)
  • 47 db(A)
  • 47 db(A)
  • Shelves/drawers:
  • 3 adjustable shelves
  • 3 adjustable shelves
  • 3 adjustable shelves
  • 3 adjustable shelves
  • 3 adjustable shelves
  • 3 adjustable shelves
  • 3 adjustable shelves
  • 3 adjustable shelves
  • 3 adjustable shelves
  • Door type:
  • 2 outer doors, 4 inner compartments
  • 2 outer doors, 4 inner compartments
  • 2 outer doors, 4 inner compartments
  • 2 outer doors, 4 inner compartments
  • 2 outer doors, 4 inner compartments
  • 2 outer doors, 4 inner compartments
  • 2 outer doors, 4 inner compartments
  • 2 outer doors, 4 inner compartments
  • 2 outer doors, 4 inner compartments
  • Door lock:
  • Yes, with key lock
  • Yes, with key lock
  • Yes, with key lock
  • Yes, with key lock
  • Yes, with key lock
  • Yes, with key lock
  • Yes, with key lock
  • Yes, with key lock
  • Yes, with key lock
  • Access ports:
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • Environmental certification:
  • EnergyStar; ISO 14001
  • EnergyStar; ISO 14001
  • EnergyStar; ISO 14001
  • EnergyStar; ISO 14001
  • EnergyStar; ISO 14001
  • EnergyStar; ISO 14001
  • EnergyStar; ISO 14001
  • EnergyStar; ISO 14001
  • EnergyStar; ISO 14001
  • Weight:
  • 288 kg (635 lbs.)
  • 226 kg (499 lbs.)
  • 288 kg (635 lbs.)
  • 226 kg (499 lbs.)
  • 288 kg (635 lbs.)
  • 247 kg (545 lbs.)
  • 247 kg (545 lbs.)
  • 288 kg (635 lbs.)
  • 247 kg (545 lbs.)
Packages with 120 V ULT freezer plus storage for 3" cryoboxes
Frequently Bought Together