CrossLab Gas Tight Syringes, Agilent Technologies
Catalog # CASG003987
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Volume:25 µl
- Description:Syringe, SGE, gas-tight
- Environmentally Preferable:
- Cat. no.:CASG003987
- Needle Length:50 mm
- Needle Type:Fixed
- Needle Gauge:23
- Tip Type:Cone
Specifications
About this item
Gas tight syringes for dispensing gases and liquids.
- Precisely made for optimal and repeatable performance through accurate volume delivery
These gas tight syringes are constructed of borosilicate glass and PTFE plunger tips. An interference fit between the plunger tip and the barrel enhances the gas sealing properties of the syringe.