Digital Ammeter, 1 - 10 A, Eisco Scientific | Avantor
Digital Ammeter, 1 - 10 A, Eisco Scientific
Digital ammeter, 0 - 10 A, dimensions: 5.5L×4.25H×4"W
Catalog # 470336-124
Supplier:   Wards
Fournisseur #:  PH1102B
Digital Ammeter, 1 - 10 A, Eisco Scientific
Catalog # 470336-124
Supplier:  Wards
Digital Ammeter, 1 - 10 A, Eisco Scientific
Digital Ammeter, 1 - 10 A, Eisco Scientific
Digital ammeter, 0 - 10 A, dimensions: 5.5L×4.25H×4"W
Price
79,55 $
Log in pour la disponibilité
Each
Product Details & Documents
Light and portable digital ammeter used to measure amperage from 0 - 10 A.
  • Light and portable with large LCD screen
  • Required 9 V battery (included)
Easy to read digital ammeter is battery operated and can be used in the classroom, lab, or in the field. Standard instrument for physics and electronic courses.
Spécifications
  • Désignation:
    Digital ammeter, 0 - 10 A, dimensions: 5.5L×4.25H×4"W