VWR® Antistatic Ionizer
Fournisseur: VWR International
Certificats
À propos de cet article
The VWR® antistatic ionizer aids in neturalizing electric charges in the air and on samples.
- Neutralizes electric charges in the environment
- Useful for measurements with non-conducting materials (plastic, glass)
- Samples can be 5 to 50 cm away from ionizer
The VWR® antistatic ionizer works by removing overflow of positive and negative ions. An inbalance of negative and positivie ions can cause repeatabilty errors during measurements, the VWR® antistatic ionizer works to neutralize the inbalance to aid measurement accuracy.
Frequently Bought Together