ESD Rubber Table Mats, Low Profile, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
The MT2500 series ESD rubber mats are made from proven 0.060" thick 2-ply rubber.
- Dissipative work surface 10E6 to 10E8 ohms Rtt
- Resistance to ground 8 × 10E6-8 ohms
- Resistance point to point 2 × 10E6-8 ohms
A conductive layer backs the static dissipative top layer that creates outstanding electrical performance characteristics. The 2500 series offers excellent resistance to hot solders, soldering irons and most solvents. MT2500 series is available in 50' rolls in 24, 30, and 36" widths. Pre-cut mat sizes are packaged individually with two (2) female 10 mm snaps and a common point ground cord.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20 and the recommendations of ESD 4.1.
Frequently Bought Together