ESD Anti-Fatigue Mats, ⅜", Transforming Technologies
VinylStat FM2 is constructed with a closed-cell expanded PVC that results in a cushioning spongy surface and provides a path-to-ground for electrostatic charges.
- ESD Anti-fatigue matting
- Brown or gray
- ⅜" thick
The mat is available in standalone workstation mats or in 60' rolls. Worker fatigue has been shown to lower productivity and morale which has a large impact on profitability. Damage due to static can have a similar drain on the bottom line - solve both problems with the VinylStat FM2 anti-fatigue matting.
Frequently Bought Together