extraktLAB Delta 8 Lab Equipment Start Up Package, United Science
This 50 L reactor is tailored for Delta 8 production. Equipment features air pumps, heater, solvent reservoirs, manual valves. Helium fluid transfer. Overhead stirrer with air motor manual system, 50 L stirred reactor, air motor, heat exchanger coupling, high temperature ultra-fast oil recirculating heater.
- Scalable industrial start up package
- Includes prepackaged consumable kit
- Low cost start up to produce approx. 500 g of Delta 8 per day from CBD isolate
This process will produce Delta 8 oils at approximately 80 to 85% potency. The oil is then remediated to remove any excess Delta 9, or is diluted to keep the Delta 9 below 0.3%.
: The Delta 8 Process and Methods Service/Commissioning provides production methods, training, and processes for Delta 8 production from CBD distillate.
Frequently Bought Together