Hook and Loop Heel Grounders, 1 Meg, Transforming Technologies
Fournisseur: Transforming Technologies
Certificats
À propos de cet article
Heel grounders connect the wearer to ground via a proper floor mat or flooring material.
- Reversible cup design for long life
- Stretch hook and loop closure strap allows maximum fit and comfort
- Available in blue and yellow
The HG1910 series Heel Grounders are constructed from durable materials and feature a stretch hook and loop closure to maximize fit and comfort. The 1" reversible cup is designed to extend the life of the device.
Frequently Bought Together