Premium Coil Cords, Transforming Technologies
A unique, flat-face snap allows for a universally solid connection to standard wristbands.
- Premium straight coil cords
- 4 mm snap
- 6, 12, and 20' lengths
The Premium Coil Cord has a 2.5 mm polyurethane coil insulation that offers excellent coil memory. A unique, flat-face snap allows for a universally solid connection to standard wrist bands. A shorter, swivel type banana jack increases coil life, prevents cord tangles and fits most bench grounds. Equipped with a 1 meg ohm resistor.
Frequently Bought Together