CC2695P Series Dual Wire Coil Cords, Transforming Technologies
Fournisseur: Transforming Technologies
Certificats
À propos de cet article
Polyurethane coil insulation and strain relief molding offers excellent coil memory and endurance.
- 'One snap' dual wire design contains two snaps in a molded head
- Innovative parking jack to hold coil cord plug
- 5, 10, and 20' lengths (extended)
- 1 meg ohm resistor
The molded snap head provides a convenient parking jack for the cord when not in use. Two female snaps, a 3.5 mm phono plug, and 1 meg resistor are standard.
Frequently Bought Together