WB0070 Dual Conductor Fabric Wrist Strap
Catalog # 76490-234
Supplier: Transforming Technologies
Specifications
- Color:Blue
- Description:Dual conductor wrist band
- Cat. no.:76490-234
Specifications
About this item
The WB0070 wrist strap features a silver-plated, monofilament, continuous thread woven together with elastic nylon for full conductivity and comfort.
- Premium one-size-fits-most adjustable fabric wrist strap
- Wrist strap makes superior contact with the skin for a reliable grounding product
- Compatible with most available resistance monitors