9010 Series ESD Jackets, Lapel Collar, Knit Cuff, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
The 9010 Series ESD jackets provide high-quality ESD protection at a great value.
- Level 3 ESD jacket
- Maintains consistent continuity after 100 wash cycles
- Snap-front closure
- Three patch pockets
The 9010 Series jackets are fabricated from lightweight 90% polyester and 10% carbon fabric for unmatched operator comfort. Jackets feature ESD knit cuffs, and lapel style collar.
Available in blue, black, white, teal, and maroon.
Specifications
- Catalog No:
- 76489-768
- 76489-814
- 76489-770
- 76489-812
- 76489-764
- 76489-810
- 76489-766
- 76489-808
- 76489-772
- 76489-774
- 76489-798
- 76489-796
- 76489-760
- 76489-806
- 76489-762
- 76489-804
- 76489-756
- 76489-802
- 76489-758
- 76489-800
- 76489-746
- 76489-790
- 76489-748
- 76489-792
- 76489-786
- 76489-788
- 76489-754
- 76489-822
- 76489-820
- 76489-750
- 76489-794
- 76489-818
- 76489-752
- 76489-816
- 76489-830
- 76489-828
- 76489-826
- 76489-824
- 76489-834
- 76489-832
- 76489-776
- 76489-782
- 76489-784
- 76489-778
- 76489-780
- Model:
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- 9010
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon
- 90% polyester and 10% carbon