Sole Grounders, 2 Meg, Transforming Technologies
Fournisseur: Transforming Technologies
Certificats
À propos de cet article
Full coverage sole grounders provide 20 times the surface contact of heel grounders.
- Wider and more consistent contact area ensures grounding
- Rapid and complete static dissipation due to constant contact through entire walking motion
ESD Sole Grounders (or ESD Foot Grounders) offer over 20 times the surface contact of standard ESD heel straps. This surface area advantage extends the life of the sole grounder over standard heel grounders, which fail over time due to even the slightest contamination, necessitating replacement or cleaning, wasting time, and costing money.
Frequently Bought Together