Electrical Connecting Cord | Avantor
Electrical Connecting Cord
Supplier:   Wards
Electrical Connecting Cord
Supplier:  Wards
Certificates
Electrical Connecting Cord
Electrical Connecting Cord
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Perfect for any application requiring basic circuit assembly.
  • 0.15" (4 mm) insulated plug on both ends
A must have in any physics classroom or laboratory.
Product Family Options
Informations sur le produit
  • Désignation
    Cord Length
    Availability
    Price
  • Electrical connecting cord, black
    500 mm (20")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.70
    $15.70
    Each
       $15.70
    Supplier Number: PH1055D-SPL1
  • Electrical connecting cord, red
    500 mm (20")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.25
    $15.25
    Each
       $15.25
    Supplier Number: PH1055T24