Surface Resistance Meter Travel Kit, Transforming Technologies
Catalog # 76490-218
Supplier: Transforming Technologies
Specifications
- Description:Surface resistance meter travel kit
- Cat. no.:76490-218
Specifications
About this item
This battery powered unit is equipped with built-in parallel electrodes for quick surface measurements.
- Easy-to-read, color-coded readout screen
- Two lightweight 'press and test' travel electrodes
- Built-in resistivity probes
- Test range: : 10³ to 10¹²
The surface resistance meter includes two lightweight 'travel' electrodes that eliminate the cumbersome weight of standard kits. When pressed, the travel electrodes simulate the 5 lbs. of pressure necessary to measure resistance point-to-point (RPP), or resistance-to-ground (RTG). This meter is designed for quick checks of surface resistance for ESD control applications in an electronics manufacturing or handling environment.