ContactUsLinkComponent
Surface Resistance Probe, 5 lbs., Transforming Technologies
  76490-216
Surface Resistance Probe, 5 lbs., Transforming Technologies
  76490-216
 :  SR0065
  • Description:
    5 lbs. Surface resistance probe
  • Cat. no.:
    76490-216
Designed to test point-to-point (RTT) and point-to-ground resistance (RTG) in accordance with ESD Association Standards.
  • 5 lbs. disk electrode probes for use with Surface Resistance Meters
  • Compatible with most resistance meters
  • Yellow PVC sleeves
The probe can be used to make measurements on floors, mats, workbenches, garments, seating, or any other flat surface where the resistance between two points is required.