ContactUsLinkComponent
Air Ionizing Gun, Transforming Technologies
Catalogue # 76489-744
Air Ionizing Gun, Transforming Technologies
Catalogue # 76489-744
Supplier Number:  IN6200
Spécifications
  • Description:
    Ac ionizing gun
  • Cat. no.:
    76489-744
Spécifications
À propos de cet article
A lightweight but durable static removal gun.
  • Point and shoot precision ionization
  • Durable impact-resistant fiberglass reinforced case
  • Quick disconnect air fittings
The ionizing gun provides superior control and targeted particulate cleanup for a wide variety of industrial applications. As a forceful neutralizer and blow-off gun, it easily removes stubborn particulate from surfaces. As a neutralizer, it gets rid of troublesome charges responsible for the attraction of contaminants to product surfaces.