Scion Series Ion Bar Display Terminal, Transforming Technologies
Catalog # 76489-742
Supplier: Transforming Technologies
Specifications
- Description:Scion series ion bar display terminal
- Cat. no.:76489-742
Specifications
About this item
View the current switching frequency and duty cycle settings and adjust them to fit specific applications.
- View current switching frequency and duty cycle settings
- Displays the bar settings
- Used with IN1200 Scion Series Ion Bars
An external display which displays the bars settings. View the current switching frequency and duty cycle settings and adjust them to fit specific applications. Without the display, a charge plate monitor is recommended to assist with the bar settings.