Disposable Heel Grounder, Transforming Technologies
Catalog # 76489-732
Supplier: Transforming Technologies
Specifications
- Color:Yellow
- Description:Disposable heel grounder
- Size:24" L
- Cat. no.:76489-732
Specifications
About this item
Disposable heel grounders are comfortable to wear, easily identifiable, and use the entire volume of the fabric to provide protection.
- Low cost method of grounding temporary personnel
- High visibility yellow color
- Disposable design is perfect for one time use
Constructed of a durable non-woven fabric impregnated with carbon, these disposable heel grounders use the entire volume of the fabric to provide protection.