Non-Marking Heel Grounder, with 35 mm Cup and Buckle Closure, Transforming Technologies
Catalogue # 76489-726
Fournisseur: Transforming Technologies
Spécifications
- Color:Red
- Description:Non-marking heel grounder with 35 mm cup and buckle closure
- Cat. no.:76489-726
Spécifications
À propos de cet article
The non-marking sport style heel grounders are ideal for boots, cross-training athletic shoes, and other odd sized shoes.
- Sport heel grounders are ideal for boots and cross-training athletic shoes
- Moldable back strap better grips shoes for a tighter fit
- Buckle closure ensures a tight and comfortable fit on the shoe
Constructed from durable materials, with a moldable backing which better grips shoes for a tighter fit, this heel grounder provides protection for staff on the move. A buckle closure makes it easy to put on and take off the device. Made with gray ESD rubber that will not leave marks on floors.