ContactUsLinkComponent
Heel Grounder, with 1.25" Cup and Buckle Closure, Transforming Technologies
  76489-724
Heel Grounder, with 1.25" Cup and Buckle Closure, Transforming Technologies
  76489-724
 :  HG1380
  • Description:
    Heel grounder with 1.25" cup and buckle closure
  • Cat. no.:
    76489-724
Constructed from durable materials with a 0.75" buckle closure to maximize fit and comfort.
  • Buckle closure
  • Buried 1 meg resistor for safety
  • Meets or exceeds requirements of ANSI ESD-S20.20
Heel grounding devices provide protection for staff on the move.