- Description:Table mat ground kit, co mmon point ground and universal snap kit
- Length:15'
- Cat. no.:76489-714
Easy to install kit for grounding ESD table mats.
- Includes 15' low profile ground cord, and a universal snap
- Universal snap kit easily installs a ground snap into a range of mat thicknesses
- Includes two screws for thick and thin materials
The CP2522 is a low profile common point ground with two banana jacks which allow two workers the ability to plug into one ground point. The molded connection point houses a 1 meg Ohm, ¹/₄ watt resistor.
The ground cord attaches to table mats via the Universal Snap Kit. The Universal Snap Kit includes a 10 mm stud, 10 mm socket, threaded backing washer, conical nut, long screw, short screw, and ESD common point ground label. Everything needed to install a stud or socket on all soft materials up to 0.375" thick. Screws are 6-32 flat head Phillips machine screws; one 0.25" long and one 0.375" long. Punch a hole through the mat and screw on the socket or stud, whichever is preferred. All that is needed is a Phillips screwdriver.