Mat Interconnect Cord, with Two 10 mm Male Snaps, Transforming Technologies
Catalog # 76489-684
Supplier: Transforming Technologies
Specifications
- Description:Mat interconnect cord, with two 10 mm male snaps
- Length:10'
- Cat. no.:76489-684
Specifications
About this item
The ground cord is used to interconnect two monitored work surfaces when unable to connect individually to ground.
- 10' length
- No resistor (zero meg)
- 10 mm male snaps
Straight cord with male snap fasteners at each end and no resistor. It is used to interconnect two monitored work surfaces together. Supplied with two molded low profile mushroom caps with ³/₈" (10 mm) male snaps to ensure a secure connection to mats.