Nomex® Heat Resistant Sleeve, Transforming Technologies
Catalog # 76489-644
Supplier: Transforming Technologies
Specifications
- Clothing Size:Universal
- Garment application:Industrial,Cleanroom,Electronics/semi-conductor
- Risk area:Heat
- Length:18"
- Cat. no.:76489-644
Specifications
About this item
Comfortable and washable, these heat-resistant sleeves protect upper arms from elevated temperatures.
- Resists heat up to 572 degrees F
- Made with Nomex® fabric
- Cleanroom compatible
- Static dissipative
Comfortable and washable, these heat-resistant sleeves protect upper arms from elevated temperature applications without compromising cleanliness or potential damage due to static discharge. The FG series Nomex® hot sleeves are designed for printed circuit board, hybrid circuit, semiconductor, and other processes that require handling objects at high temperatures. They fit easily over most gloves.