ContactUsLinkComponent
Socket Connection, 10 mm to 10 mm, Transforming Technologies
  76489-638
undefined
Socket Connection, 10 mm to 10 mm, Transforming Technologies
  76489-638
 :  CS0128-SOCKET
  • Description:
    10 mm socket to socket connection
  • Cat. no.:
    76489-638
This Snap Adapter Set is compatible and interchangeable with all major snap manufactures.
  • Steel adapter has a double 10 mm stud construction
  • Used to convert 10 mm snaps, primarily with anti-static matting and ground cords
  • Snap adapter connects a 10 mm socket to a 10 mm socket
Also available in a stud-to-stud connection with the CS0128-stud.