ContactUsLinkComponent
Stud Connection, 10 mm to 10 mm, Transforming Technologies
Stud Connection, 10 mm to 10 mm, Transforming Technologies
Catalog # 76489-636
undefined
Stud Connection, 10 mm to 10 mm, Transforming Technologies
Catalog # 76489-636
Supplier Number:  CS0128
Specifications
  • Description:
    10 mm stud to stud connection
  • Cat. no.:
    76489-636
Specifications
About this item
This Snap Adapter Set is compatible and interchangeable with all major snap manufactures.
  • Steel adapter has a double 10mm socket construction
  • Used to convert 10 mm snaps, primarily with anti-static matting and ground cords
  • Snap adapter connects a 10 mm stud to a 10 mm stud
Also available in a socket-to-socket connection with the CS0128-socket.