Molded Banana Jack Snap, 10 mm Stud and Socket, Transforming Technologies
Molded banana jack snap, 10 mm stud and socket
Catalog # 76489-634
Supplier: Transforming Technologies
Fournisseur #: CS0127
Molded Banana Jack Snap, 10 mm Stud and Socket, Transforming Technologies
Catalog # 76489-634
Supplier: Transforming Technologies
Jump to:
Product Details & Documents
The Molded Banana Jack Snap adds a banana jack to a table mat.
- Add a banana jack to any surface with a 10 mm ground snap
- Molded plastic body with metal snaps
- Use with coil cords, wrist strap sets, common point grounds, and constant monitors
Fully molded in plastic, the device has both male and female 10 mm snaps with a banana jack, and is compatible and interchangeable with standard banana plugs. It can connect to a wrist strap, ground cord, or constant monitor, and is excellent for grounding carts.
Spécifications
- Désignation:Molded banana jack snap, 10 mm stud and socket