Interconnect Cord, with 2 Banana Plugs, Transforming Technologies
Catalog # 76489-586
Supplier: Transforming Technologies
Specifications
- Description:Interconnect cord, with 2 banana plugs
- Length:8'
- Cat. no.:76489-586
Specifications
About this item
Two standard banana plugs allow the cord to connect two devices such as mats or ground cords that feature banana jacks.
- Use to connect two items together
- Each end terminates with a standard banana plug
- Length is 8'
- No resistor
The Interconnect Cord features 2.5 mm polyurethane insulation that offers excellent cord life.