Wolfgang Warmbier Two Point Probe, Transforming Technologies
Catalog # 76489-572
Supplier: Transforming Technologies
Specifications
- Description:SRM mini probe, model 410, for resistance testers SRM110 and SRM200
- Cat. no.:76489-572
Specifications
About this item
The Two-Point Probe is designed to measure surface resistance of small areas.
- Measures point-to-point surface resistance of small areas
- Spring-loaded contact pins
- Connects to meters with banana plugs
- Works with the SRM110 and SRM200 surface resistance meters
The probe works specifically with the SRM110 and SRM200 surface resistance meters, and may work with other models as well.