Wolfgang Warmbier PGT120 ESD Footwear Tester, Transforming Technologies
76489-564
- Description:ESD footwear tester
- Cat. no.:76489-564
The personnel grounding tester is able to perform a quick and accurate control measurement of the ESD protection. The test results are indicated both with audible and visual signals. The PGT120 measures the left and right shoe separately.
- Independently test the functioning of ESD footwear and wrist strap grounding products and signals
- Automatically test footwear in hands-free mode
- Audibly and visually signal a 'Pass', 'High-Fail', or 'Low-Fail'
- Tests both feet simultaneously and independently to save time
The PGT120 is an easy to use and complete test station for ESD shoes and wrist straps. Test results are indicated both with audible and visual signals. The PGT120 measures the left and right shoe separately.