Conductive Trash Can Dolly, Transforming Technologies
Catalog # 76490-264
Supplier: Transforming Technologies
Specifications
- Description:Conductive trash dolly
- Cat. no.:76490-264
Specifications
About this item
The cart provides a user-friendly solution for heavy ESD trash containers.
- Industrial aluminum construction
- Wider than plastic dolly
- Solid aluminum hubs
The dolly locks on to the 44 gallon WBAS180 Transforming Technologies container. The 5 heavy-duty casters provide stability for loads up to 350 lbs., and two casters are conductive to provide a path to ground for ESD charges.