Cove N Curve™ Mop Adapter, Micronova
Supplier: Micronova
Certificates
About this item
This uniquely shaped mop adapter transforms a traditional flat mop design into a curved cleaning surface. Designed to address coved joints, exterior of pipes, the interior of drums and small tanks, as well as other curved surfaces.
- Constructed from PTFE with a Polyketone swivel joint
- Swivel joint allows an easy glide
- ACME thread adapter
- Resistant to common cleaning chemicals and disinfectants
- Available in 9" or 16" (23 or 41 cm) lengths
- Autoclavable
The Micronova Cove N Curve™ Mop Adapter is designed for use with SnapMop™ and PocketMop™ series flat head mops and connects directly to the STA series threaded handles, fixed length or extendible.
Handles
Product Information
Frequently Bought Together