Ladle, Long Handle, Disposable, Bürkle
Supplier: BURKLE INC
Certificates
About this item
The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates.
- PS white, PS blue or Bio-PE
- Individually packaged and optionally sterilized
- Reaches into any corner
- Handle length 300 mm (14.37")
- For single use
- Clean room manufactured (clean room class 7)
The ladle has a very smooth bottom and rounded corners. This makes it possible to reach into any corner of a container or to take medium directly from the bottom of the container. A long handle with indentations for fingers ensures a firm hold and precise work. The disposable ladle can be used in the pharmaceutical industry, chemistry and cosmetics and the food and animal feed industries.
Certifications: Complies with EU food requirements and FDA regulations.
Frequently Bought Together