Level 2 Easy-Breathe SMS Lab Coats, Apex Aseptic Products
Easy-breathe lab coats are manufactured from AAMI Level 2 SMS fabrics.
- Made with AAMI level 2 SMS fabrics
- Fluid resistant and easy breathability
- Hydrohead fluid resistant test: 229 mm
- Spray impact test: 0.83 g
- Low particle shedding; category 1 per Helmke Drum test
- Manufactured in controlled environment
Easy-breathe SMS lab coats are latex-free and nonsterile. These knee length garments feature a snap-front closure, 3 pockets, knit cuffs, and a knit collar. Available in a wide variety of colors and sizes.
: Manufacturing facility is ISO 9001 and ISO 13485 certified. FDA registered and inspected.
: Lab coats are packaged 10 per pack, 100 per case.
- Catalog No:
- 76488-706
- 76488-708
- 76488-746
- 76488-702
- 76488-748
- 76488-704
- 76488-734
- 76488-736
- 76488-730
- 76488-732
- 76488-742
- 76488-700
- 76488-744
- 76488-738
- 76488-740
- 76488-728
- 76488-724
- 76488-726
- 76488-750
- 76488-712
- 76488-756
- 76488-714
- 76488-758
- 76488-752
- 76488-754
- 76488-710
- 76488-720
- 76488-722
- 76488-716
- 76488-718
- Fabric:
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- SMS
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Pockets:
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- With pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
Frequently Bought Together