About this item
All New Zealand calf serum products are carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases. We follow the same standards in processing of our calf serum as with our Fetal Bovine Serum (FBS) to enable offering the highest quality and traceability.
- A cost-effective alternative to Fetal Bovine Serum (FBS)
- Sourced from New Zealand with complete traceability back to original source
- Age of animal at time of collection less than 10 days old
- Low in antibodies and high in growth factors
- Virus panel testing according to 9 CFR 113.53
New Zealand Newborn Bovine Calf Serum is filtered through sequential 100 nm (0.1 µm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76324-936
- CA76324-904
- CA76324-934
- CA76324-902
- Origin:
- Animal
- Animal
- Animal
- Animal
- Endotoxin:
- ≤25 EU/ml
- ≤25 EU/ml
- ≤25 EU/ml
- ≤25 EU/ml
- Hemoglobin:
- ≤30 mg/dl
- ≤30 mg/dl
- ≤30 mg/dl
- ≤30 mg/dl
- Storage temperature:
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.