The animals are fed an enriched diet to ensure proper nutrition and are fasted prior to bleeding to reduce lipid concentration. The horses are bled in an enclosed facility that is specially equipped for aseptic collection. Coggins tests for equine infectious anemia are routinely performed on each horse.
- Tested for equine infectious anemia
- Complete traceability back to original source
- Virus panel testing according to 9 CFR 113.53
onor Equine Serum is filtered through three sequential 100 nm (0,1 μm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- CA76534-630
- CA76534-632
- CA76324-874
- CA76534-628
- CA82013-588
- CA76236-290
- Origin:
- US
- US
- US
- US
- US
- US
- Endotoxin:
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- Hemoglobin:
- ≤10 mg/dl
- ≤10 mg/dl
- ≤10 mg/dl
- ≤10 mg/dl
- ≤10 mg/dl
- ≤10 mg/dl
- Storage temperature:
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.