Whirl-Pak® Sterilized Write-On Bags, Nasco Sampling
À propos de cet article
Blended, virgin polyethylene film with Write-On strip provides exceptional clarity, superior strength, and sterilized after manufacturing for results you can trust. HACCP, EPA, USDA, FDA approved for food contact.
- Plus 1 sterilization, post-manufacturing sterilization: Increased sample integrity, perforated closure to maintain sterility
- Convenient write-on strip for easy labeling
- Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
- Opening pull tabs: For easy opening, non-slip textured Tyvek
- Continuous perimeter seal: Maximum protection against leaks
- Tape extends beyond wire to eliminate sharp ends and provide a sturdy grip, wire tabs hold the top open for easy access to add to or remove sample, worry-free, no-leak closure for sample integrity
Bags should not be used at temperatures above 180 °F (82 °C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to –346 °F (–210 °C) —careful handling is required after freezing. Capacity is measured when the bag is closed and tab is folded over three times. Volume and dimensions are approximate.
Shelf life from sterilization date found on the sterility documentation: 5 years from sterilized date found on your sterility confirmation.
Certifications: Approved by USDA, FDA, EPA and HACCP. Food safe.
7 Options Available Below
Product Details & Documents
Blended, virgin polyethylene film with Write-On strip provides exceptional clarity, superior strength, and sterilized after manufacturing for results you can trust. HACCP, EPA, USDA, FDA approved for food contact.
- Plus 1 sterilization, post-manufacturing sterilization: Increased sample integrity, perforated closure to maintain sterility
- Convenient write-on strip for easy labeling
- Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
- Opening pull tabs: For easy opening, non-slip textured Tyvek
- Continuous perimeter seal: Maximum protection against leaks
- Tape extends beyond wire to eliminate sharp ends and provide a sturdy grip, wire tabs hold the top open for easy access to add to or remove sample, worry-free, no-leak closure for sample integrity
Bags should not be used at temperatures above 180 °F (82 °C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to –346 °F (–210 °C) —careful handling is required after freezing. Capacity is measured when the bag is closed and tab is folded over three times. Volume and dimensions are approximate.
Shelf life from sterilization date found on the sterility documentation: 5 years from sterilized date found on your sterility confirmation.
Certifications: Approved by USDA, FDA, EPA and HACCP. Food safe.
Documents
Product Family Options
Informations sur le produit
- DésignationCapacitéÉpaisseurW×LAvailabilityPrice
- Round wire29 ml (1 oz.)2.25 mil.64×127 mm (2.5×5")Discontinued-Supplier Number: -
- Round wire29 ml (1 oz.)2.25 mil.64×127 mm (2.5×5")Discontinued-Supplier Number: -
- Round wire59 ml (2 oz.)2.25 mil.76×127 mm (3×5")Log in pour la disponibilitéCase of 10000Case of 10000 - $7,316.93$7,316.93Prix promotionnel pour les nouveaux clientsEach$365.85Pack of 500$370.62Case of 10000$7,316.93Supplier Number: B01064
- Round wire118 ml (4 oz.)2.25 mil.76×184 mm (3×7.25")Log in pour la disponibilitéCase of 10000Case of 10000 - $6,557.44$6,557.44Prix promotionnel pour les nouveaux clientsEach$327.87Pack of 500$376.09Case of 10000$6,557.44Supplier Number: B01062
- Round wire384 ml (13 oz.)2.5 mil.130×190 mm (5.12×7.48")Discontinued-Supplier Number: -
- Round wire532 ml (18 oz.)2.25 mil.114×229 mm (4.5×9")Log in pour la disponibilitéCase of 6000Case of 6000 - $5,411.65$5,411.65Prix promotionnel pour les nouveaux clientsEach$450.97Pack of 500$529.33Case of 6000$5,411.65Supplier Number: B01065
- Round wire710 ml (24 oz.)3 mil.152×229 mm (6×9")Log in pour la disponibilitéPack of 500Pack of 500 - $869.24$869.24Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 3000Case of 3000 - $5,215.46$5,215.46Prix promotionnel pour les nouveaux clientsPack of 500$869.24Case of 3000$5,215.46Supplier Number: B01297
- 1 flat, 1 round wire2720 ml (92 oz.)4 mil.254×381 mm (10×15")Log in pour la disponibilitéCase of 500Case of 500 - $805.74$805.74Prix promotionnel pour les nouveaux clientsEach$402.87Pack of 250$472.88Case of 500$805.74Supplier Number: B01445
- 1 flat, 1 round wire3637 ml (123 oz.)4 mil.254×508 mm (10×20")Log in pour la disponibilitéBox of 250Box of 250 - $1,095.10$1,095.10Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéEachEach - $1,095.10$1,095.10Prix promotionnel pour les nouveaux clientsBox of 250$1,095.10Each$1,095.10Supplier Number: B01446
- 1 flat, 1 round wire5441 ml (184 oz.)4 mil.381×508 mm (15×20")Log in pour la disponibilitéPack of 100Pack of 100 - $348.55$348.55Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 200Case of 200 - $697.10$697.10Prix promotionnel pour les nouveaux clientsPack of 100$348.55Case of 200$697.10Supplier Number: B01447
- Discontinued-Supplier Number: -