Blue Biopsy Wraps
À propos de cet article
Mopec’s Blue Biopsy Wraps are high-quality, disposable biopsy specimen wraps designed to securely hold and protect small tissue samples during laboratory processing. They are made from strong blue lens paper that contrasts with tissue samples, making it easier for histology technicians to locate tiny biopsies throughout processing and embedding.
Specimen protection: wraps help contain and maintain the identity of small biopsy tissue during handling, transfer, and processing, reducing the risk of loss or damage
- High visibility: blue color provides excellent contrast against most tissue types and paraffin, aiding quick visual identification during embedding and sectioning
- Chemical resistance: lens paper construction resists common histology reagents such as water, alcohol, and xylene, ensuring the wrap doesn’t deteriorate or interfere as specimens move through processing steps
- Flexible sizing: multiple sizes allow labs to choose the best fit for different biopsy dimensions, improving efficiency and reducing waste
These wraps are available in multiple sizes (including 2.5×4″, 2×3″, 2×2.5″, and 1.5×2″) to accommodate a range of specimen sizes and preferences in workflow.
Mopec’s Blue Biopsy Wraps stand out for their flexible sizing, reagent resistance, and contrast visibility, making them a solid, cost-effective choice in many histology labs. These wraps are widely used in pathology labs to support accurate specimen tracking and handling in routine histological workflows.
4 Options Available Below
Product Details & Documents
Mopec’s Blue Biopsy Wraps are high-quality, disposable biopsy specimen wraps designed to securely hold and protect small tissue samples during laboratory processing. They are made from strong blue lens paper that contrasts with tissue samples, making it easier for histology technicians to locate tiny biopsies throughout processing and embedding.
Specimen protection: wraps help contain and maintain the identity of small biopsy tissue during handling, transfer, and processing, reducing the risk of loss or damage
- High visibility: blue color provides excellent contrast against most tissue types and paraffin, aiding quick visual identification during embedding and sectioning
- Chemical resistance: lens paper construction resists common histology reagents such as water, alcohol, and xylene, ensuring the wrap doesn’t deteriorate or interfere as specimens move through processing steps
- Flexible sizing: multiple sizes allow labs to choose the best fit for different biopsy dimensions, improving efficiency and reducing waste
These wraps are available in multiple sizes (including 2.5×4″, 2×3″, 2×2.5″, and 1.5×2″) to accommodate a range of specimen sizes and preferences in workflow.
Mopec’s Blue Biopsy Wraps stand out for their flexible sizing, reagent resistance, and contrast visibility, making them a solid, cost-effective choice in many histology labs. These wraps are widely used in pathology labs to support accurate specimen tracking and handling in routine histological workflows.
Product Family Options
Informations sur le produit
- DimensionsPackagingColorAvailabilityPrice
- 1.5×2"500 papers/boxBlueEach$79.77Pack of 500$55.14Supplier Number: BI057
- 2×2.5"500 papers/boxBlueEach$82.81Pack of 500$57.67Supplier Number: BI058
- 2×3"500 papers/boxBlueEach$122.86Pack of 500$87.85Supplier Number: BI059
- 2.5×4"500 papers/boxBlueEach$122.86Pack of 500$87.85Supplier Number: BI060