Formalin Neutralizer Pads, Mortech® | Avantor
Formalin Neutralizer Pads, Mortech®
Supplier:   Mopec
Formalin Neutralizer Pads, Mortech®
Supplier:  Mopec
Certificates
Formalin Neutralizer Pads, Mortech®
Formalin Neutralizer Pads, Mortech®
3 Options Available Below
Product Details & Documents
Dramatically minimizes exposure to formaldehyde vapors.
  • Minimizes cleanup
  • Incineration of pads is clean and efficient with minimal threat of exposure
  • Fiber absorbs and then locks in liquids so that work areas stay drier
  • Resists tearing, even when saturated
  • Internal sliding shelf for easy positioning cube for pouring and storage
  • Simple and easy to use
A major crisis facing today's hospitals, laboratories and other healthcare providers is the occupational exposure and disposal of formaldehyde, a known carcinogen. Formaldehyde Neutralizer Pads offer a way to significantly reduce exposure by chemically neutralizing formaldehyde and encapsulating the escaping vapors with the pad.
Clinical Pad applications include surgical cutting board pads, dissecting pads, autopsy table pads, and liners for body trays and cadaver bags. Formaldehyde Neutralizer Pads reduce exposure by chemically bonding with formaldehyde, resulting in the encapsulation of vapors-ideal for grossing areas and autopsy work.
Product Family Options
Informations sur le produit
  • Taille
    Dimensions
    Availability
    Price
  • Small
    8×11"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $325.71
    $325.71
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $195.84
    $195.84
    Each
       $325.71
    Pack of 25
       $195.84
    Supplier Number: BE093
  • Medium
    11×15"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $522.47
    $522.47
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $346.57
    $346.57
    Each
       $522.47
    Pack of 25
       $346.57
    Supplier Number: BE094
  • Large
    15×20"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $651.58
    $651.58
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $462.15
    $462.15
    Each
       $651.58
    Pack of 25
       $462.15
    Supplier Number: BE095