ContactUsLinkComponent
Eazy Soak, Instrument Cleaner, Mortech®
Eazy Soak, Instrument Cleaner, Mortech®
Catalog # 76498-464
Supplier:  Mopec
undefined
Eazy Soak, Instrument Cleaner, Mortech®
Catalog # 76498-464
Supplier:  Mopec
Supplier Number:  BE046
Specifications
  • Description:
    Eazy soak, instrument cleaner
  • Size:
    32 oz.
  • Cat. no.:
    76498-464
Specifications
About this item
This unique, clear, odorless enzyme instrument cleaner is ideal for an overnight easy soak.
  • No scrubbing or pre-rinsing is required
  • 32 oz. container
Ideal for hard-to-clean instruments such as scissors, forceps, serrated cutting blades, and scalpel handles. It digests all tissue, blood, and exudative material.