ContactUsLinkComponent
Bone Dust Collector Hood for Mopec Swordfish, Mortech®
Catalogue # 76498-410
Fournisseur:  Mortech Manufacturing
undefined
Bone Dust Collector Hood for Mopec Swordfish, Mortech®
Catalogue # 76498-410
Fournisseur:  Mortech Manufacturing
Supplier Number:  BD147
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Connector hood
  • Cat. no.:
    76498-410
Spécifications
À propos de cet article
Disposable hoods for the Mopec Swordfish Bone Dust Collector system.
  • Flexible PVC hoods
  • Simple to install over the already mounted blades
Flexible PVC hoods include the hose hook-up for the vacuum system. The bone dust collector hoods will work with all Mopec Swordfish blades, except for the 76 mm large round blade (76498-404).
Packaging: Sold in packs of 10.