Double Dial Hanging Autopsy Scale, Mortech®
Fournisseur: Mortech Manufacturing
Certificats
À propos de cet article
Heavy chrome scale body and pan suspension.
- Accurate to 0.1% of full scale
- Double 13" dial (read from either side)
Oversized air dashpots to absorb shock and bring to a quick stop.
Delivery: Bow, ring, and pan included with set.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together