Coplin Staining Jar, Mortech® | Avantor
Coplin Staining Jar, Mortech®
Supplier:   Mopec
Coplin Staining Jar, Mortech®
Supplier:  Mopec
Certificates
Coplin Staining Jar, Mortech®
Coplin Staining Jar, Mortech®
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Heavy, clear glass jar with four molded internal grooves to hold up to ten slides.
  • Glass base construction
  • Four molded grooves in glass hold up to ten 3×1" slides
Jar features a broad base to prevent spillage. Both glass and plastic lid options protect against contaminants. The plastic option is a tight-fitting screw lid to prevent fluid evaporation. The glass cover option simply and easily sets upon the jar.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Staining jar with glass lid
    110H×16Ø mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $71.29
    $71.29
    Each
       $71.29
    Supplier Number: BA002
  • Staining jar with plastic lid
    110H×16Ø mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $71.29
    $71.29
    Each
       $71.29
    Supplier Number: BA003