Borosil® Solution Bottle Borosilicate Glass, Plain Tooled Neck, Foxx Life Sciences®
Supplier: Foxx Life Sciences
Borosil® Solution Bottle Borosilicate Glass, Plain Tooled Neck, Foxx Life Sciences®
Supplier: Foxx Life Sciences
Certificates
Jump to:
About this item
Borosilicate glass 3.3 bottles, with tooled neck, high mechanical strength and chemical resistant.
- Bottles are specially designed for handling and storage of culture media
- Matt-finished enameled area for marking
- Mechanically strong and chemical resistant
- Round, no cap
Borosil laboratory bottles are chemically resistant and stable. Fitted with a tooled neck. The bottles are autoclavable and sterilizable.
9 Options Available Below
Product Details & Documents
Borosilicate glass 3.3 bottles, with tooled neck, high mechanical strength and chemical resistant.
- Bottles are specially designed for handling and storage of culture media
- Matt-finished enameled area for marking
- Mechanically strong and chemical resistant
- Round, no cap
Borosil laboratory bottles are chemically resistant and stable. Fitted with a tooled neck. The bottles are autoclavable and sterilizable.
Product Family Options
Product Information
- DésignationCapacitéØHauteurAvailabilityPrice
- Borosil® solution bottle borosilicate glass, plain tooled neck250 ml70 mm140 mmLog in for availabilityCase of 20$616.60Supplier Number: 1585021
- Borosil® solution bottle borosilicate glass, plain tooled neck500 ml80 mm180 mmLog in for availabilityCase of 20$768.01Supplier Number: 1585024
- Borosil® solution bottle borosilicate glass, plain tooled neck1 L (1000 ml)101 mm230 mmLog in for availabilityCase of 10$700.76Supplier Number: 1585029
- Borosil® solution bottle borosilicate glass, plain tooled neck2 L (2000 ml)136 mm285 mmLog in for availabilityCase of 10$997.80Supplier Number: 1585030
- Borosil® solution bottle borosilicate glass, plain tooled neck3 L (3000 ml)156 mm255 mmLog in for availabilityCase of 4$654.80Supplier Number: 1585031
- Borosil® solution bottle borosilicate glass, plain tooled neck5 L (5000 ml)183 mm350 mmLog in for availabilityEach$497.08Supplier Number: 1585033
- Borosil® solution bottle borosilicate glass, plain tooled neck10 L (10000 ml)235 mm430 mmLog in for availabilityEach$618.97Supplier Number: 1585038
- Borosil® solution bottle borosilicate glass, plain tooled neck20 L (20000 ml)256 mm530 mmLog in for availabilityEach$1,283.04Supplier Number: 1585040
- Borosil® solution bottle borosilicate glass, plain tooled neck50 L (50000 ml)417 mm665 mmLog in for availabilityEach$7,803.23Supplier Number: 1585046