Borosil® Reagent Square Bottles GL 80 PP Screw Cap PP Pouring Ring, Foxx Life Sciences®
Borosil® Laboratory bottles are chemically resistant and stable.
- Mechanically strong and chemical resistant
- Designed with plastic pouring ring for drip-free operation
- Screw caps and pouring ring are made of PP
- Polypropylene pour ring
When fitted with a plastic pouring ring dripping can be totally eliminated. As there is only one size of screw thread for all bottles, the screw caps and pouring rings are fully interchangeable. The bottles, pouring rings and caps are autoclavable and sterilizable.
: IS 1388 (Part II), EN ISO 4796-2.
Frequently Bought Together