Sterile DuraGrip Tyvek™ Knee-High Boot Covers, Apex Aseptic Products
Fournisseur: APEX ASEPTIC PRODUCTS, LLC.
Certificats
À propos de cet article
Sterile DuraGrip Dupont® Tyvek™ knee-high boot covers are suitable for cleanroom use.
- Fluid resistant yet breathable
- Low particle shedding, rated category 1 per Helmke Drum Test
- Latex-free, TSE-free, BSE-free
- Manufactured in a controlled environment
- Gamma irradiated validated sterile to SAL 10⁻⁶
- Suitable to use in ISO Class 5 or higher cleanrooms
Boot cover soles are made with specially engineered DuraGrip PVC material, which is ultra-non-skid and durable. The upper part of the boot cover is made with Dupont® Tyvek™ fabrics. Tunneled elastic sewing around the ankle and knee reduces particle shedding. The garments are specially folded to ensure proper aseptic donning.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 Certified and FDA registered and inspected.
Delivery: Each package includes a gamma irradiation indicator dot, and each shipping case is affixed with a Certificate of Conformance and Certificate of Sterility.
Packaging: Boot covers are packaged 2 pieces per package, triple bagged, 200 pieces per case.
Frequently Bought Together