VWR® REDISHIP General Purpose Fume Hoods with Cupsink, Kewaunee Scientific
Supplier: VWR International
Certificates
About this item
General purpose fume hoods that meet the typical needs of a research environment, while maintaining a level of flexibility. Ready for quick delivery through the REDISHIP program.
- LED lighting
- Pre-wired fan switch
- Alarm cutout with cover
- Stainless steel duct collar
- 120 VAC GFI duplex on each post
- stainless steel air foil frameless laminated safety glass sash
- Choice of 4, 5 or 6' width
Hoods includes Kemglass liner stainless steel. Fit for 48, 60 or 72" black Kemresin fume hood worktop (must be ordered separately).
Certifications: UL / CUL labeled.
Ordering information: Choice of 48, 60 or 72" wide Black Kemresin (chemical resistant epoxy resin) fume hood worktops. These include left rear cupsink cutout and cover, right rear acid vent hole with cover. Worktops must be ordered separately.
Specifications
- Catalog No:
- KS-05G5472LED
- KS-05G5448LED
- KS-05G5460LED
- Sash opening:
- 28” max
- 28” max
- 28” max
- Dimensions:
- 72×36×54”
- 48×36×54”
- 60×36×54”